b-AP15 [DUB inhibitor] solution 98%

Supplier: Ambeed
Warning

Synonyms: VLX1500, (3E,5E)-1-Acryloyl-3,5-bis(4-nitrobenzylidene)piperidin-4-one

A619221-10MG A619221-1MG A619221-25MG A619221-5MG
77157-718EA 66.95 USD
77157-718 77157-720 77157-722 77157-724
b-AP15 [DUB inhibitor] solution 98%
b-AP15 [DUB inhibitor] solution

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃O₆
MW: 419.4 g/mol
MDL Number: MFCD26142662
CAS Number: 1009817-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR