2-(4-Bromophenyl)-1,3-dioxolane 95%
Supplier: Ambeed
|
Warning
|
A618378-25G
A618378-5G
A618378-1G
77157-708EA
175
USD
77157-708
77157-710
77893-244
2-(4-Bromophenyl)-1,3-dioxolane 95%
2-(4-Bromophenyl)-1,3-dioxolane
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ |
MDL Number:
MFCD01318950 CAS Number: 10602-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...