8-Fluoroquinoline-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A614923-100MG A614923-250MG
77157-694EA 156.56 USD
77157-694 77157-696
8-Fluoroquinoline-4-carbaldehyde 95%
8-Fluoroquinoline-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆FNO
MW: 175.15 g/mol
MDL Number: MFCD06824407
CAS Number: 496792-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR