Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97%

Supplier: Ambeed
Warning

Synonyms: 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane

A612665-100G A612665-25G A612665-5G
77157-682EA 199 USD
77157-682 77157-684 77157-686
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97%
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane

Precaución: for Research Use Only

Formula: C₁₂H₁₉F₉O₃Si
MDL Number: MFCD09954454
CAS Number: 102390-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR