(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-(tert-butoxycarbonyl)piperidin-4-yl)propanoic acid 98%

Supplier: Ambeed
Warning

A611993-1G A611993-250MG A611993-5G A611993-25G A611993-100MG
77157-676EA 115 USD
77157-676 77157-678 77157-680 77572-994 77893-235
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-(tert-butoxycarbonyl)piperidin-4-yl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-(tert-butoxycarbonyl)piperidin-4-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₄N₂O₆
MDL Number: MFCD01321423
CAS Number: 204058-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR