2,5-Difluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A607655-100MG A607655-1G A607655-250MG
77157-668EA 8 USD
77157-668 77157-670 77157-672
2,5-Difluorobenzeneboronic acid pinacol ester 98%
2,5-Difluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₂O₂
MDL Number: MFCD12405346
CAS Number: 408492-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR