Bis(NHS)PEG5 98%

Supplier: Ambeed
Warning

Synonyms: 4,7,10,13,16-Pentaoxanonadecanedioic Acid Di(N-succinimidyl) Ester, Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16-pentaoxanonadecane-1,19-dioate, Di(N-succinimidyl) 4,7,10,13,16-Pentaoxanonadecanedioate

A607408-1G A607408-100MG A607408-250MG A607408-5G
77157-666EA 132 USD
77157-666 77551-822 77686-533 77686-534
Bis(NHS)PEG5 98%
Bis(NHS)PEG5

Precaución: for Research Use Only

Formula: C₂₂H₃₂N₂O₁₃
MDL Number: MFCD11041118
CAS Number: 756526-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR