(±)-trans-1,2-Diaminocyclohexane 96%

Supplier: Ambeed
Danger

Synonyms: (±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine

A606472-100G A606472-10G A606472-25G A606472-5G
77157-658EA 64.89 USD
77157-658 77157-660 77157-662 77157-664
(±)-trans-1,2-Diaminocyclohexane 96%
(±)-trans-1,2-Diaminocyclohexane

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00063747
CAS Number: 1121-22-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR