WS3 98%

Supplier: Ambeed
Warning

Synonyms: N-(6-(4-(3-(4-((4-Methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)ureido)phenoxy)pyrimidin-4-yl)cyclopropanecarboxamide, N-[6-[4-[[[[4-[(4-Methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-amino]carbonyl]amino]phenoxy]-4-pyrimidinyl]-cyclopropanecarboxamid

A606355-10MG A606355-1MG A606355-25MG A606355-50MG
77157-650EA 47.38 USD
77157-650 77157-652 77157-654 77157-656
WS3 98%
WS3

Precaución: for Research Use Only

Formula: C₂₈H₃₀F₃N₇O₃
MW: 569.589 g/mol
MDL Number: MFCD26142950
CAS Number: 1421227-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR