4-Methoxy-3-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A606068-1G A606068-250MG A606068-5G
77157-644EA 10 USD
77157-644 77157-646 77157-648
4-Methoxy-3-methylbenzonitrile 98%
4-Methoxy-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD12797513
CAS Number: 53078-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR