trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A604687-100MG A604687-1G A604687-250MG A604687-5G
77157-632EA 7 USD
77157-632 77900-975 77901-087 77900-841
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate 97%
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆FNO₃
MDL Number: MFCD18791200
CAS Number: 869481-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR