TWS119 98%

Supplier: Ambeed
Warning

Synonyms: 3-[[6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]phenol, 3-((6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenol

A604058-10MG A604058-1G A604058-1MG A604058-25MG A604058-5MG
77157-616EA 126.69 USD
77157-616 77157-618 77157-620 77157-622 77157-624
TWS119 98%
TWS119

Precaución: for Research Use Only

Formula: C₁₈H₁₄N₄O₂
MW: 318.34 g/mol
MDL Number: MFCD09037535
CAS Number: 601514-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR