4'-Bromo-2',5'-difluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-2,5-difluorophenyl)ethanone

A603685-1G A603685-250MG A603685-5G
77157-610EA 46.35 USD
77157-610 77157-612 77157-614
4'-Bromo-2',5'-difluoroacetophenone 97%
4'-Bromo-2',5'-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.0255 g/mol
MDL Number: MFCD15526896
CAS Number: 123942-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR