Ethyl 2-(1H-benzo[d]imidazol-2-yl)acetate 98%

Supplier: Ambeed
Warning

A602453-1G A602453-5G
77157-596EA 14 USD
77157-596 77157-598
Ethyl 2-(1H-benzo[d]imidazol-2-yl)acetate 98%
Ethyl 2-(1H-benzo[d]imidazol-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.22 g/mol
MDL Number: MFCD00624872
CAS Number: 14741-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR