2,8-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzo[b,d]furan 95%

Supplier: Ambeed
Warning

A601541-100MG A601541-1G A601541-250MG
77157-584EA 16.48 USD
77157-584 77157-586 77157-588
2,8-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzo[b,d]furan 95%
2,8-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₂₄H₃₀B₂O₅
MW: 420.11 g/mol
MDL Number: MFCD28396385
CAS Number: 1197989-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR