4-Chloro-2,6-difluoronitrobenzene 95%

Supplier: Ambeed
Warning

A599907-10G A599907-1G A599907-250MG A599907-25G A599907-5G
77157-574EA 195 USD
77157-574 77157-576 77157-578 77157-580 77157-582
4-Chloro-2,6-difluoronitrobenzene 95%
4-Chloro-2,6-difluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MDL Number: MFCD20039793
CAS Number: 136272-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR