3-Methyl-2-oxazolidone 98%

Supplier: Ambeed
Warning

A599246-100G A599246-25G A599246-5G
77157-568EA 82 USD
77157-568 77157-570 77900-667
3-Methyl-2-oxazolidone 98%
3-Methyl-2-oxazolidone

Precaución: for Research Use Only

Formula: C₄H₇NO₂
MDL Number: MFCD00009759
CAS Number: 19836-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR