Methyl 5-chloro-2-hydroxy-4-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

A595288-1G A595288-250MG
77157-550EA 287.37 USD
77157-550 77157-552
Methyl 5-chloro-2-hydroxy-4-(trifluoromethyl)benzoate 95%
Methyl 5-chloro-2-hydroxy-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
MDL Number: MFCD28405105
CAS Number: 1103837-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR