Hydroxyfasudil Hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-((1,4-Diazepan-1-yl)sulphonyl)isoquinolin-1(2H)-one hydrochloride, Hydroxyfasudil HCl, 4-diazepan-1-ylsulphonyl)-2H-isoquinolin-1-one, 5-(1, hydrochloride, HA-1100 hydrochloride

A591593-1MG A591593-5MG
77157-534EA 32.96 USD
77157-534 77157-536
Hydroxyfasudil Hydrochloride 98%
Hydroxyfasudil Hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClN₃O₃S
MW: 343.83 g/mol
MDL Number: MFCD06411567
CAS Number: 155558-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR