N-Ethyl-o-crotonotoluidide 98%

Supplier: Ambeed
Warning

Synonyms: (E)-N-Ethyl-N-(o-tolyl)but-2-enamide, N-Crotonyl-N-ethyl-o-toluidine, trans-N-Ethyl-N-(o-tolyl)-2-butenamide

A591274-25G A591274-5G
77157-528EA 24.72 USD
77157-528 77157-530
N-Ethyl-o-crotonotoluidide 98%
N-Ethyl-o-crotonotoluidide

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO
MW: 203.29 g/mol
MDL Number: MFCD00026989
CAS Number: 124236-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR