1,4-Dimethyl-1,2,4-triazolium iodide 97%

Supplier: Ambeed
Danger

A591077-250MG A591077-100MG A591077-5G
77157-522EA 28 USD
77157-522 77902-045 77900-718
1,4-Dimethyl-1,2,4-triazolium iodide 97%
1,4-Dimethyl-1,2,4-triazolium iodide

Precaución: for Research Use Only

Formula: C₄H₈IN₃
MDL Number: MFCD00666522
CAS Number: 120317-69-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR