(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 1-(6-amino-3,5-difluoropyridin-2-yl)-8-chloro-6-fluoro-7-(3-hydroxyazetidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Delafloxacin meglumine
A587080-10MG
A587080-1MG
A587080-50MG
A587080-5MG
77157-496EA
16.48
USD
77157-496
77157-498
77157-500
77157-502
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 1-(6-amino-3,5-difluoropyridin-2-yl)-8-chloro-6-fluoro-7-(3-hydroxyazetidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylate 98%
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 1-(6-amino-3,5-difluoropyridin-2-yl)-8-chloro-6-fluoro-7-(3-hydroxyazetidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₂₅H₂₉ClF₃N₅O₉ MW: 635.97 g/mol |
MDL Number:
MFCD26142921 CAS Number: 352458-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...