1-Boc-3-ethynyl-3-hydroxyazetidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-ethynyl-3-hydroxyazetidine-1-carboxylate

A586786-1G A586786-250MG
77157-492EA 310.03 USD
77157-492 77157-494
1-Boc-3-ethynyl-3-hydroxyazetidine 98%
1-Boc-3-ethynyl-3-hydroxyazetidine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
MDL Number: MFCD21324699
CAS Number: 1259034-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR