3-Chloro-5-iodopyridin-2-ol 95+%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-iodo-pyridin-2-ol

A586424-100MG A586424-1G
77157-474EA 28.84 USD
77157-474 77157-476
3-Chloro-5-iodopyridin-2-ol 95+%
3-Chloro-5-iodopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃ClINO
MW: 255.44 g/mol
MDL Number: MFCD11227187
CAS Number: 97966-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR