Diglycolic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Oxydiacetic acid

A585759-25G A585759-5G A585759-100G A585759-500G
77157-470EA 19.57 USD
77157-470 77157-472 77594-774 77686-485
Diglycolic acid 98%
Diglycolic acid

Precaución: for Research Use Only

Formula: C₄H₆O₅
MW: 134.09 g/mol
MDL Number: MFCD00004309
CAS Number: 110-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR