(5-Nitro-1H-indol-1-yl)acetic acid 95+%

Supplier: Ambeed
Warning

A584150-1G A584150-250MG A584150-5G
77157-446EA 55.62 USD
77157-446 77157-448 77157-450
(5-Nitro-1H-indol-1-yl)acetic acid 95+%
(5-Nitro-1H-indol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.18 g/mol
MDL Number: MFCD11205787
CAS Number: 226901-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR