(3,4-Difluoro-5-hydroxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A583113-100MG A583113-1G A583113-250MG
77157-432EA 35.02 USD
77157-432 77157-434 77157-436
(3,4-Difluoro-5-hydroxyphenyl)boronic acid 98%
(3,4-Difluoro-5-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD22414421
CAS Number: 1379466-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR