4-Bromo-2-fluoro-5-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A582191-100MG A582191-250MG
77157-428EA 30.9 USD
77157-428 77157-430
4-Bromo-2-fluoro-5-methoxybenzaldehyde 98%
4-Bromo-2-fluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
MDL Number: MFCD17012743
CAS Number: 749931-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR