(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline 95%

Supplier: Ambeed
Danger

Synonyms: (R,R)-QuinoxP*, 2,3-Bis((R)-tert-butyl(methyl)phosphino)quinoxaline

A578539-100MG A578539-1G A578539-250MG A578539-5G
77157-414EA 70.04 USD
77157-414 77157-416 77157-418 77686-471
(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline 95%
(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂P₂
MW: 334.38 g/mol
MDL Number: MFCD10565649
CAS Number: 866081-62-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR