CID-2011756 98+%

Supplier: Ambeed
Warning

Synonyms: 5-(3-Chlorophenyl)-N-[4-(4-morpholinylmethyl)phenyl]-2-furancarboxamide, 5-(3-Chlorophenyl)-N-(4-(morpholinomethyl)phenyl)furan-2-carboxamide, 5-(3-Chloro-phenyl)-furan-2-carboxylic acid (4-morpholin-4-ylmethyl-phenyl)-amide

A576631-10MG A576631-1MG A576631-50MG A576631-5MG
77157-400EA 39.14 USD
77157-400 77157-402 77157-404 77157-406
CID-2011756 98+%
CID-2011756

Precaución: for Research Use Only

Formula: C₂₂H₂₁ClN₂O₃
MW: 396.87 g/mol
MDL Number: MFCD04094273
CAS Number: 638156-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR