Potassium 3,5-dibromophenyltrifluoroborate 95+%

Supplier: Ambeed
Warning

A576514-1G A576514-250MG A576514-5G
77157-394EA 20 USD
77157-394 77157-396 77157-398
Potassium 3,5-dibromophenyltrifluoroborate 95+%
Potassium 3,5-dibromophenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BBr₂F₃K
MDL Number: MFCD26383469
CAS Number: 929626-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR