GSK-3787 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-N-(2-((5-(trifluoromethyl)pyridin-2-yl)sulphonyl)ethyl)benzamide, GSK3787

A575893-10MG A575893-1MG A575893-50MG A575893-5MG
77157-386EA 93 USD
77157-386 77157-388 77157-390 77157-392
GSK-3787 98%
GSK-3787

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClF₃N₂O₃S
MDL Number: MFCD00099612
CAS Number: 188591-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR