9-Chloro-5,6,7,8-tetrahydroacridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A575699-100MG A575699-250MG A575699-50MG
77157-380EA 183.34 USD
77157-380 77157-382 77157-384
9-Chloro-5,6,7,8-tetrahydroacridine-3-carboxylic acid 95%
9-Chloro-5,6,7,8-tetrahydroacridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClNO₂
MW: 261.70 g/mol
MDL Number: MFCD14779907
CAS Number: 902586-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR