3-Bromo-4-fluorobenzylamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A575490-1G
A575490-25G
A575490-5G
77157-374EA
14
USD
77157-374
77157-376
77157-378
3-Bromo-4-fluorobenzylamine hydrochloride 98%
3-Bromo-4-fluorobenzylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₈BrClFN |
MDL Number:
MFCD00143094 CAS Number: 202865-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...