6-Bromo-4-chloropyridine-2,3-diamine 95%
Supplier: Ambeed
|
Warning
|
A574373-100MG
A574373-250MG
77157-362EA
94.76
USD
77157-362
77157-364
6-Bromo-4-chloropyridine-2,3-diamine 95%
6-Bromo-4-chloropyridine-2,3-diamine
Precaución: for Research Use Only
Formula:
C₅H₅BrClN₃ MW: 222.47 g/mol |
MDL Number:
MFCD29044935 CAS Number: 1841081-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...