2-(4-(1,1-Difluoroethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Danger

Synonyms: 1,3,2-Dioxaborolane, 2-[4-(1,1-difluoroethyl)phenyl]-4,4,5,5-tetramethyl-

A572801-100MG A572801-1G A572801-250MG A572801-5G
77157-348EA 36.05 USD
77157-348 77157-350 77157-352 77157-354
2-(4-(1,1-Difluoroethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-(1,1-Difluoroethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₉BF₂O₂
MW: 268.10 g/mol
MDL Number: MFCD13186728
CAS Number: 1000994-94-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR