Fmoc-D-Lys(Ivdde)-OH 97%

Supplier: Ambeed
Warning

A572332-1G A572332-250MG A572332-5G A572332-25G
77157-342EA 106 USD
77157-342 77157-344 77157-346 77569-096
Fmoc-D-Lys(Ivdde)-OH 97%
Fmoc-D-Lys(Ivdde)-OH

Precaución: For research use only.

Formula: C₃₄H₄₂N₂O₆
MDL Number: MFCD06796894
CAS Number: 1272755-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR