(2S,3R)-Allyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Thr-OAll

A571645-100MG A571645-1G A571645-250MG A571645-5G
77157-336EA 18 USD
77157-336 77157-338 77157-340 77900-756
(2S,3R)-Allyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate 95%
(2S,3R)-Allyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₅
MDL Number: MFCD23160326
CAS Number: 136523-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR