(3-Methyl-4-morpholinophenyl)boronic acid 97%

Supplier: Ambeed
Warning

A571539-1G A571539-250MG
77157-332EA 63.86 USD
77157-332 77157-334
(3-Methyl-4-morpholinophenyl)boronic acid 97%
(3-Methyl-4-morpholinophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD16495874
CAS Number: 1426245-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR