6-Bromo-3-chloro-2-cyanopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-chloropicolinonitrile

A571229-100MG A571229-250MG
77157-328EA 39.14 USD
77157-328 77157-330
6-Bromo-3-chloro-2-cyanopyridine 98%
6-Bromo-3-chloro-2-cyanopyridine

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.46 g/mol
MDL Number: MFCD17014673
CAS Number: 1252046-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR