4-(5-Fluoro-2-nitrophenyl)morpholine 97%

Supplier: Ambeed
Warning

A570203-100MG A570203-1G A570203-250MG
77157-316EA 19.57 USD
77157-316 77157-318 77157-320
4-(5-Fluoro-2-nitrophenyl)morpholine 97%
4-(5-Fluoro-2-nitrophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₁FN₂O₃
MDL Number: MFCD09834357
CAS Number: 536977-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR