BMN-673 98%

Supplier: Ambeed
Warning

Synonyms: (8R,9S)-5-fluoro-8-(4-fluorophenyl)-9-(1-methyl-1H-1,2,4-triazol-5-yl)-8,9-dihydro-1H-pyrido[4,3,2-de]phthalazin-3(2H)-one, Talazoparib, (8S,9R)-5-Fluoro-8-(4-fluorophenyl)-9-(1-methyl-1H-1,2,4-triazol-5-yl)-8,9-dihydro-2H-pyrido[4,3,2-de]phthalazin-3(7H)-one

A570059-10MG A570059-1MG A570059-250MG A570059-2MG A570059-50MG A570059-5MG A570059-1G
77157-298EA 46.35 USD
77157-298 77157-300 77157-302 77157-304 77157-306 77157-308 77686-453
BMN-673 98%
BMN-673

Precaución: for Research Use Only

Formula: C₁₉H₁₄F₂N₆O
MW: 380.35 g/mol
MDL Number: MFCD22666357
CAS Number: 1207456-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR