Ethyl 4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate

A569752-100G A569752-25G A569752-1000G A569752-500G
77157-294EA 8.24 USD
77157-294 77157-296 77613-780 77613-782
Ethyl 4-hydroxybenzoate 98%
Ethyl 4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002353
CAS Number: 120-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR