N-Boc benzylamine 95%

Supplier: Ambeed
Warning

A569714-100G A569714-25G
77157-292EA 272.95 USD
77157-292 77572-814
N-Boc benzylamine 95%
N-Boc benzylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.26 g/mol
MDL Number: MFCD05148681
CAS Number: 42116-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR