3-(Thiophen-3-yl)propan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-Thienyl)-1-propanol

A569109-100MG A569109-1G A569109-250MG
77157-286EA 27 USD
77157-286 77157-288 77157-290
3-(Thiophen-3-yl)propan-1-ol 98%
3-(Thiophen-3-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₇H₁₀OS
MDL Number: MFCD09927266
CAS Number: 20905-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR