5-Bromo-2-fluoro-3-methoxypyridine 98%

Supplier: Ambeed
Warning

A566760-100MG A566760-250MG A566760-25G A566760-5G A566760-1G
77157-274EA 7 USD
77157-274 77157-276 77157-278 77157-280 77686-443
5-Bromo-2-fluoro-3-methoxypyridine 98%
5-Bromo-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD09909847
CAS Number: 880870-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR