3'-Chloro-5'-(trifluoromethoxy)acetophenone 95+%

Supplier: Ambeed
Warning

A566096-1G A566096-250MG A566096-5G
77157-264EA 27.81 USD
77157-264 77157-266 77157-268
3'-Chloro-5'-(trifluoromethoxy)acetophenone 95+%
3'-Chloro-5'-(trifluoromethoxy)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.59 g/mol
MDL Number: MFCD06660315
CAS Number: 886503-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR