8-(5-(5,7-Dihydroxy-4-oxo-4H-chromen-2-yl)-2-methoxyphenyl)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

A561680-1MG A561680-10MG
77157-254EA 37 USD
77157-254 77893-142
8-(5-(5,7-Dihydroxy-4-oxo-4H-chromen-2-yl)-2-methoxyphenyl)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one 95%
8-(5-(5,7-Dihydroxy-4-oxo-4H-chromen-2-yl)-2-methoxyphenyl)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₃₂H₂₂O₁₀
MDL Number: MFCD00597035
CAS Number: 548-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR