(S)-N-Fmoc-α-methyl-2-fluorophenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)-2-methylpropanoic acid, FMOC-α-ME-L-PHE(2-F)-OH, FMOC-α-METHYL-D-2-FLUOROPHENYLALANINE

A560713-100MG A560713-1G A560713-250MG A560713-5G
77157-246EA 48.41 USD
77157-246 77157-248 77157-250 77157-252
(S)-N-Fmoc-α-methyl-2-fluorophenylalanine 95%
(S)-N-Fmoc-alpha-methyl-2-fluorophenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₂FNO₄
MW: 419.452 g/mol
MDL Number: MFCD17019320
CAS Number: 1172127-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR