AZ-5104 98+%

Supplier: Ambeed
Warning

Synonyms: N-(5-((4-(1H-Indol-3-yl)pyrimidin-2-yl)amino)-2-((2-(dimethylamino)ethyl)(methyl)amino)-4-methoxyphenyl)acrylamide

A557620-10MG A557620-1MG A557620-50MG A557620-5MG
77157-220EA 40.17 USD
77157-220 77157-222 77157-224 77157-226
AZ-5104 98+%
AZ-5104

Precaución: for Research Use Only

Formula: C₂₇H₃₁N₇O₂
MW: 485.58 g/mol
MDL Number: MFCD28899992
CAS Number: 1421373-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR